2,2,6-Trimethyl-4H-1,3-dioxin-4-one ≥94% contains up to 6% acetone

Supplier: BeanTown Chemical

126855-100G 126855-25G 126855-500G
BT126855-100GEA 81 USD
BT126855-100G BT126855-25G BT126855-500G
2,2,6-Trimethyl-4H-1,3-dioxin-4-one ≥94% contains up to 6% acetone
2,2,6-Trimethyl-4H-1,3-dioxin-4-one

CAS: 5394-63-8; EC No: 226-403-3; MDL No: MFCD00040468 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C7H10O3; MW: 142.15 Melting Point: 12-13°; Boiling Point: 65-67°/2 mmHg; Flash point: 14°C (57°F) Density (g/mL): 1.07; Refractive Index: 1.460


  • Refractive index: 1.460
  • Liquid

Formula: C₇H₁₀O₃
Boiling Pt: 65…67 °C/2 mmHg
Melting Pt: 12…13 °C
Density: 1.07 g/mL
Flash Pt: 14 °C (57 °F)
MDL Number: MFCD00040468
CAS Number: 5394-63-8
EINECS: 226-403-3
ADR: 3,II

Order Now

Specification Test Results

Assay 94%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR