Thulium ≥99.9% (trace rare earth metals basis), pieces, sublimed dendritic
Supplier: BeanTown Chemical
126840-1G
126840-5G
BT126840-1GEA
88
USD
BT126840-1G
BT126840-5G
Thulium ≥99.9% (trace rare earth metals basis), pieces, sublimed dendritic
Thulium
CAS: 7440-30-4; EC No: 231-140-2; MDL No: MFCD00011281
UN No: UN3178; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: Tm; MW: 168.93
Melting Point: 1545°; Boiling Point: 1950°
Density (g/mL): 9.332
- Solid
Formula:
Tm MW: 168.93 g/mol Boiling Pt: 1950 °C Melting Pt: 1545 °C Density: 9.332 g/mL |
MDL Number:
MFCD00011281 CAS Number: 7440-30-4 EINECS: 231-140-2 UN: 3178 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...