5-Acetylvaleric acid ≥96%
Supplier: BeanTown Chemical
126640-10G
126640-50G
BT126640-10GEA
117.51
USD
BT126640-10G
BT126640-50G
5-Acetylvaleric acid ≥96%
5-Acetylvaleric acid
CAS: 3128-07-2; EC No: 221-512-2; MDL No: MFCD00051479
UN No: UN3261; Haz Class: 8; Packing Group: III
Solid; Molecular Formula: C7H12O3; MW: 144.17
Melting Point: 35-37°; Boiling Point: 158-162°/9 mmHg
Density (g/mL): 1.059
- Solid
Formula:
C₇H₁₂O₃ Boiling Pt: 158…162 °C/9 mmHg Melting Pt: 35…37 °C Density: 1.059 g/mL |
MDL Number:
MFCD00051479 CAS Number: 3128-07-2 EINECS: 221-512-2 ADR: 8,III |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...