1,3-Diphenyl-2-thiourea ≥98%
Supplier: BeanTown Chemical
Synonyms:
Thiocarbanilide, N,N'-Diphenylthiourea, N,N’-Diphenylthiourea
126620-100G
126620-2.5KG
126620-500G
BT126620-100GEA
20.36
USD
BT126620-100G
BT126620-2.5KG
BT126620-500G
1,3-Diphenyl-2-thiourea ≥98%
1,3-Diphenyl-2-thiourea
CAS: 102-08-9; MDL No: MFCD00004921; RTECS: FE1225000
UN No: UN2811; Haz Class: 6.1; Packing Group: II
Powder; Linear Formula: (C6H5NH)2CS; Molecular Formula: C13H12N2S ; MW: 228.32
Melting Point: 150-154°
Density (g/mL): 1.32
- RTECS: FE1225000
- Powder
Formula:
C₁₃H₁₂N₂S MW: 228.32 g/mol Melting Pt: 150…154 °C Density: 1.32 g/mL |
MDL Number:
MFCD00004921 CAS Number: 102-08-9 UN: 2811 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...