Magnesium ethanolate
Supplier: BeanTown Chemical
Synonyms:
Magnesium ethoxide, Magnesium ethylate
126565-1KG
126565-250G
126565-50G
BT126565-1KGEA
318
USD
BT126565-1KG
BT126565-250G
BT126565-50G
Magnesium ethanolate
Magnesium ethanolate
CAS: 2414-98-4; EC No: 219-323-5; MDL No: MFCD00009060
UN No: UN3205; Haz Class: 4.2; Packing Group: II
Granular; Linear Formula: Mg(OCH2CH3)2; MW: 114.44
Melting Point: 270° (decomposes)
Density (g/mL): 1.01
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Granular
Formula:
C₄H₁₀MgO₂ MW: 114.43 g/mol Melting Pt: 270 °C (dec.) Density: 1.01 g/mL |
MDL Number:
MFCD00009060 CAS Number: 2414-98-4 EINECS: 219-323-5 UN: 3205 ADR: 4.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...