2-Pentene cis- and trans-mixture ≥98%
Supplier: BeanTown Chemical
126510-100G
126510-25G
BT126510-100GEA
207
USD
BT126510-100G
BT126510-25G
2-Pentene cis- and trans-mixture ≥98%
2-Pentene
CAS: 109-68-2; EC No: 203-695-0; MDL No: MFCD00009384
UN No: UN3295; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2CH=CHCH3; Molecular Formula: C5H10; MW: 70.13
Boiling Point: 37°; Flash point: -45°C (-49°F)
Density (g/mL): 0.65; Refractive Index: 1.380
- Refractive index: 1.380
- Liquid
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 37 Melting Pt: -140 Density: 0.65 g/mL Flash Pt: –45 °C (–49 °F) |
MDL Number:
MFCD00009384 CAS Number: 109-68-2 EINECS: 203-695-0 UN: 3295 ADR: 3 II Merck Index: 12,07263 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...