Gold(I) iodide ≥99%

Supplier: BeanTown Chemical

126470-1G 126470-5G
BT126470-1GEA 183.65 USD
BT126470-1G BT126470-5G
Gold(I) iodide ≥99%
Gold(I) iodide

CAS: 10294-31-2; EC No: 233-656-3; MDL No: MFCD00014175 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Molecular Formula: AuI; MW: 323.87 Melting Point: 120° (decomposes) Density (g/mL): 8.25 Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: AuI
MW: 323.87 g/mol
Melting Pt: 120 °C (dec.)
Density: 8.25 g/mL
MDL Number: MFCD00014175
CAS Number: 10294-31-2
EINECS: 233-656-3
UN: 3260
ADR: 8 III
Merck Index: 12,04530

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR