Lithium fluoride ≥99.95%

Supplier: BeanTown Chemical

126460-100G 126460-25G
BT126460-100GEA 336 USD
BT126460-100G BT126460-25G
Lithium fluoride ≥99.95%
Lithium fluoride

CAS: 7789-24-4; EC No: 232-152-0; MDL No: MFCD00011090; RTECS: OJ6125000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: LiF; MW: 25.94 Melting Point: 845°; Boiling Point: 1673° Density (g/mL): 2.64


  • RTECS: OJ6125000
  • Powder

Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1676
Melting Pt: 870
Density: 2.64 g/mL
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1 III
Merck Index: 13,05553

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR