Osmium tetraoxide 2% (w/w) in water
Supplier: BeanTown Chemical
126455-10ML
126455-2ML
BT126455-10MLEA
116.95
USD
BT126455-10ML
BT126455-2ML
Osmium tetraoxide 2% (w/w) in water
Osmium tetraoxide
CAS: 20816-12-0; EC No: 244-058-7; MDL No: MFCD00011150
UN No: UN3287; Haz Class: 6.1; Packing Group: III
Liquid; Linear Formula: OsO4; MW: 254.20
Melting Point: 0°; Boiling Point: 100°
Density (g/mL): 1.04
- Liquid
Formula:
O₄Os Boiling Pt: 100 °C Melting Pt: 0 °C Density: 1.04 g/mL |
MDL Number:
MFCD00011150 CAS Number: 20816-12-0 EINECS: 244-058-7 UN: 3287 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...