Ruthenium(III) bromide

Supplier: BeanTown Chemical

126440-1G 126440-5G
BT126440-1GEA 92 USD
BT126440-1G BT126440-5G
Ruthenium(III) bromide
Ruthenium(III) bromide

CAS: 14014-88-1; EC No: 237-829-4; MDL No: MFCD00016314 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: RuBr3; MW: 340.78 Density (g/mL): 5.3 Hygroscopic


  • Hygroscopic
  • Powder

Formula: RuBr₃
MW: 340.78 g/mol
Density: 5.3 g/mL
MDL Number: MFCD00016314
CAS Number: 14014-88-1
EINECS: 237-829-4
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR