Nickel(II) phosphate hydrate
Supplier: BeanTown Chemical
126430-1KG
126430-250G
BT126430-1KGEA
480
USD
BT126430-1KG
BT126430-250G
Nickel(II) phosphate hydrate
Nickel(II) phosphate hydrate
CAS: 27176-17-6; MDL No: MFCD00049501
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder/Lump; Linear Formula: Ni3(PO4)2·xH2O; MW: 366.01 (anhydrous)
- Powder/Lump
Formula:
Ni₃(PO₄)₂·xH₂O |
MDL Number:
MFCD00049501 CAS Number: 27176-17-6 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...