Osmium ≥99.8% (trace metals basis), powder
Supplier: BeanTown Chemical
126390-1G
126390-5G
BT126390-1GEA
130.25
USD
BT126390-1G
BT126390-5G
Osmium ≥99.8% (trace metals basis), powder
Osmium
CAS: 7440-04-2; EC No: 231-114-0; MDL No: MFCD00011147; RTECS: RN1100000
UN No: UN3089; Haz Class: 4.1; Packing Group: II
Powder; Molecular Formula: Os; MW: 190.23
Melting Point: 3045°; Boiling Point: 5027°
Density (g/mL): 22.61
- RTECS: RN1100000
- Powder
Formula:
Os Boiling Pt: 5027 °C Melting Pt: 3045 °C Density: 22.61 g/mL |
MDL Number:
MFCD00011147 CAS Number: 7440-04-2 EINECS: 231-114-0 ADR: 4.1,II |
Specification Test Results
Assay | 99.8% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...