Trimethyl orthoformate ≥99%

Supplier: BeanTown Chemical

126370-100ML 126370-2.5L 126370-500ML
BT126370-100MLEA 35.54 USD
BT126370-100ML BT126370-2.5L BT126370-500ML
Trimethyl orthoformate ≥99%
Trimethyl orthoformate

CAS: 149-73-5; EC No: 205-745-7; MDL No: MFCD00008483; RTECS: RM6650000 UN No: UN3272; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH(OCH3)3; Molecular Formula: C4H10O3; MW: 106.12 Melting Point: -57°; Boiling Point: 101-102°; Flash point: 9°C (48°F) Density (g/mL): 0.968; Refractive Index: 1.379 Moisture Sensitive


  • RTECS: RM6650000
  • Refractive index: 1.379
  • Moisture Sensitive
  • Liquid

Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 101…102 °C
Melting Pt: –57 °C
Density: 0.968 g/mL
Flash Pt: 9 °C (48 °F)
MDL Number: MFCD00008483
CAS Number: 149-73-5
EINECS: 205-745-7
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR