4-Amino-2,2-difluoro-1,3-benzodioxole ≥97%
Supplier: BeanTown Chemical
126340-1G
126340-5G
BT126340-1GEA
91
USD
BT126340-1G
BT126340-5G
4-Amino-2,2-difluoro-1,3-benzodioxole ≥97%
4-Amino-2,2-difluoro-1,3-benzodioxole
CAS: 106876-54-4; MDL No: MFCD00792416
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C7H5F2NO2 ; MW: 173.12
- Liquid
Formula:
C₇H₅F₂NO₂ |
MDL Number:
MFCD00792416 CAS Number: 106876-54-4 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...