Ethyl acrylate ≥99% stabilized
Supplier: BeanTown Chemical
Synonyms:
Acrylic acid-ethyl ester
126320-100ML
126320-2.5L
126320-500ML
BT126320-100MLEA
25.57
USD
BT126320-100ML
BT126320-2.5L
BT126320-500ML
Ethyl acrylate ≥99% stabilized
Ethyl acrylate
CAS: 140-88-5; EC No: 205-438-8; MDL No: MFCD00009188; RTECS: AT0700000
UN No: UN1917; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH2=CHCOOCH2CH3; Molecular Formula: C5H8O2; MW: 100.12
Melting Point: -71°; Boiling Point: 99°; Flash point: 8°C (46°F)
Density (g/mL): 0.918; Refractive Index: 1.406
- RTECS: AT0700000
- Refractive index: 1.406
- Liquid
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 99 °C Melting Pt: –71 °C Density: 0.918 g/mL Flash Pt: 8 °C (46 °F) |
MDL Number:
MFCD00009188 CAS Number: 140-88-5 EINECS: 205-438-8 UN: 1917 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...