3-Aminopropionitrile ≥98% stabilized
Supplier: BeanTown Chemical
Synonyms:
β-Aminopropionitrile, 3-Aminopropanenitrile
126255-1G
126255-25G
126255-5G
BT126255-1GEA
37
USD
BT126255-1G
BT126255-25G
BT126255-5G
3-Aminopropionitrile ≥98% stabilized
3-Aminopropionitrile
CAS: 151-18-8; EC No: 205-786-0; MDL No: MFCD00014820; RTECS: UG0350000
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: H2NCH2CH2CN; Molecular Formula: C3H6N2; MW: 70.10
Density (g/mL): 0.952
- RTECS: UG0350000
- Liquid
Formula:
C₃H₆N₂ MW: 70.10 g/mol Density: 0.952 g/mL |
MDL Number:
MFCD00014820 CAS Number: 151-18-8 EINECS: 205-786-0 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...