Ethyl 2-bromoisobutyrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl α-bromoisobutyrate, Ethyl 2-bromo-2-methylpropionate, 2-Brom-2-methylpropionic acid ethyl ester

126180-250G 126180-50G
BT126180-250GEA 76 USD
BT126180-250G BT126180-50G
Ethyl 2-bromoisobutyrate ≥98%
Ethyl 2-bromoisobutyrate

CAS: 600-00-0; EC No: 209-980-6; MDL No: MFCD00000123 UN No: UN2924; Haz Class: 3(8); Packing Group: III Liquid; Linear Formula: (CH3)2CBrCOOCH2CH3; Molecular Formula: C6H11BrO2; MW: 195.06 Boiling Point: 65-67°/11 mmHg; Flash point: 60°C (140°F) Density (g/mL): 1.329; Refractive Index: 1.444


  • Refractive index: 1.444
  • Liquid

Formula: C₆H₁₁BrO₂
Boiling Pt: 65…67 °C/11 mmHg
Density: 1.329 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00000123
CAS Number: 600-00-0
EINECS: 209-980-6
ADR: 3(8),III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR