Sodium tetraphenylborate ≥99%

Supplier: BeanTown Chemical

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

126160-100G 126160-25G 126160-5G
BT126160-100GEA 345.28 USD
BT126160-100G BT126160-25G BT126160-5G
Sodium tetraphenylborate ≥99%
Sodium tetraphenylborate

CAS: 143-66-8; EC No: 205-605-5; MDL No: MFCD00011494; RTECS: ED3362500 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Linear Formula: (C6H5)4BNa; Molecular Formula: C24H20BNa ; MW: 342.23 Melting Point: 300° Light Sensitive


  • RTECS: ED3362500
  • Light Sensitive
  • Solid

Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00011494
CAS Number: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR