p-Anisidine ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline

126155-100G 126155-1KG 126155-500G
BT126155-100GEA 25.68 USD
BT126155-100G BT126155-1KG BT126155-500G
p-Anisidine ≥99%
p-Anisidine

CAS: 104-94-9; EC No: 203-254-2; MDL No: MFCD00007864; RTECS: BZ5450000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Linear Formula: CH3OC6H4NH2; Molecular Formula: C7H9NO; MW: 123.16 Melting Point: 56-59°; Boiling Point: 240-243°; Flash point: 122°C (252°F) Density (g/mL): 1.089 Air Sensitive, Moisture Sensitive


  • RTECS: BZ5450000
  • Air Sensitive, Moisture Sensitive
  • Solid

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 240…243 °C
Melting Pt: 56…59 °C
Density: 1.089 g/mL
Flash Pt: 122 °C (252 °F)
MDL Number: MFCD00007864
CAS Number: 104-94-9
EINECS: 203-254-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR