tert-Butyl trichloroacetimidate ≥97%

Supplier: BeanTown Chemical

Synonyms: tert-Butyl 2,2,2-trichloroacetimidate, t-Butyl trichioroacetimidate, t-butyl-2,2,2-trichloroacetimidate

126095-1G 126095-25G 126095-5G
BT126095-1GEA 24 USD
BT126095-1G BT126095-25G BT126095-5G
tert-Butyl trichloroacetimidate ≥97%
tert-Butyl trichloroacetimidate

CAS: 98946-18-0; MDL No: MFCD00077410 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CCl3CNHOC(CH3)3; Molecular Formula: C6H10Cl3NO; MW: 218.51 Melting Point: 20-22°; Boiling Point: 65°/11 mmHg; Flash point: 55°C (131°F) Density (g/mL): 1.221; Refractive Index: 1.456 Moisture Sensitive


  • Refractive index: 1.456
  • Moisture Sensitive
  • Liquid

Formula: C₆H₁₀Cl₃NO
MW: 218.502 g/mol
Boiling Pt: 65 °C/11 mmHg
Melting Pt: 20…22 °C
Density: 1.221 g/mL
Flash Pt: 55 °C (131 °F)
MDL Number: MFCD00077410
CAS Number: 98946-18-0
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR