Copper selenide ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Copper(II) selenide
126090-100G
126090-25G
126090-5G
BT126090-100GEA
1126
USD
BT126090-100G
BT126090-25G
BT126090-5G
Copper selenide ≥99.5% (trace metals basis)
Copper selenide
CAS: 1317-41-5; EC No: 215-272-8; MDL No: MFCD00016058
UN No: UN3077; Haz Class: 9; Packing Group: III
6mm & Down Pieces; Molecular Formula: CuSe; MW: 142.50
Melting Point: 550° (decomposes)
- 6mm & Down Pieces
Formula:
CuSe MW: 142.51 g/mol Melting Pt: 550 °C (dec.) |
MDL Number:
MFCD00016058 CAS Number: 1317-41-5 EINECS: 215-272-8 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...