Bis(2-chloroethyl)ether ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Chloroethyl ether

126060-1KG 126060-250G 126060-50G
BT126060-1KGEA 397 USD
BT126060-1KG BT126060-250G BT126060-50G
Bis(2-chloroethyl)ether ≥99%
Bis(2-chloroethyl)ether

CAS: 111-44-4; EC No: 203-870-1; MDL No: MFCD00000975; RTECS: KN0875000 UN No: UN1916; Haz Class: 6.1 (3); Packing Group: II Liquid; Linear Formula: ClCH2CH2OCH2CH2Cl; Molecular Formula: C4H8Cl2O ; MW: 143.01 Melting Point: -47°; Boiling Point: 65-67°/15 mmHg; Flash point: 55°C (131°F) Density (g/mL): 1.22; Refractive Index: 1.456


  • RTECS: KN0875000
  • Refractive index: 1.456
  • Liquid

Formula: C₄H₈Cl₂O
MW: 143.01 g/mol
Boiling Pt: 178
Melting Pt: -44
Density: 1.22 g/mL
Flash Pt: 55 °C (131 °F)
MDL Number: MFCD00000975
CAS Number: 111-44-4
EINECS: 203-870-1
UN: 1916
ADR: 6.1 II
Merck Index: 13,03091

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR