Silver(I) hexafluorophosphate ≥98%

Supplier: BeanTown Chemical

Synonyms: Silver hexafluorophosphate

126045-1G 126045-25G 126045-5G
BT126045-1GEA 39.97 USD
BT126045-1G BT126045-25G BT126045-5G
Silver(I) hexafluorophosphate ≥98%
Silver(I) hexafluorophosphate

CAS: 26042-63-7; EC No: 247-428-6; MDL No: MFCD00003415 UN No: UN3260; Haz Class: 8; Packing Group: II Powder; Linear Formula: AgPF6; MW: 252.83 Melting Point: 102° (decomposes) Light Sensitive


  • Light Sensitive
  • Powder

Formula: AgF₆P
MW: 252.83
Melting Pt: 102 °C (dec.)
MDL Number: MFCD00003415
CAS Number: 26042-63-7
EINECS: 247-428-6
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR