3-Bromobenzylamine ≥97%

Supplier: BeanTown Chemical

Synonyms: (3-Bromophenyl)methanamine

126035-100G 126035-25G 126035-5G
BT126035-100GEA 524 USD
BT126035-100G BT126035-25G BT126035-5G
3-Bromobenzylamine ≥97%
3-Bromobenzylamine

CAS: 10269-01-9; MDL No: MFCD01026119 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C7H8BrN; MW: 186.05


  • Liquid

Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD01026119
CAS Number: 10269-01-9
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR