Gallium ≥99.99999% (trace metals basis), pellets, Diameter 6 mm

Supplier: BeanTown Chemical

126015-10G 126015-1G 126015-50G
BT126015-10GEA 100 USD
BT126015-10G BT126015-1G BT126015-50G
Gallium ≥99.99999% (trace metals basis), pellets, Diameter 6 mm
Gallium

CAS: 7440-55-3; EC No: 231-163-8; MDL No: MFCD00134045; RTECS: LW8600000 UN No: UN2803; Haz Class: 8; Packing Group: III Low Melting Solid; Molecular Formula: Ga; MW: 69.72 Melting Point: 29.8°; Boiling Point: 2403° Density (g/mL): 5.904


  • RTECS: LW8600000
  • Low Melting Solid

Formula: Ga
MW: 69.72 g/mol
Boiling Pt: 2403 °C
Melting Pt: 29.8 °C
Density: 5.904 g/mL
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III

Order Now

Specification Test Results

Assay 99.99999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR