Methyl toluene-4-sulfonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid methyl ester, Methyl p-toluenesulphonate, Toluene-4-sulphonic acid methyl ester

125950-1KG 125950-250G
BT125950-1KGEA 138 USD
BT125950-1KG BT125950-250G
Methyl toluene-4-sulfonate ≥98%
Methyl toluene-4-sulphonate

CAS: 80-48-8; EC No: 201-283-5; MDL No: MFCD00008417; RTECS: XT7000000 UN No: UN1759; Haz Class: 8; Packing Group: III Solid; Linear Formula: CH3C6H4SO3CH3; Molecular Formula: C8H10O3S; MW: 186.23 Melting Point: 25-28°; Boiling Point: 144-145°/5 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.234; Refractive Index: 1.517 Moisture Sensitive


  • RTECS: XT7000000
  • Refractive index: 1.517
  • Moisture Sensitive
  • Solid

Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
Boiling Pt: 144…145 °C/5 mmHg
Melting Pt: 25…28 °C
Density: 1.234 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00008417
CAS Number: 80-48-8
EINECS: 201-283-5
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR