Diphenyl diselenide ≥98%

Supplier: BeanTown Chemical

125940-100G 125940-25G 125940-5G
BT125940-100GEA 522 USD
BT125940-100G BT125940-25G BT125940-5G
Diphenyl diselenide ≥98%
Diphenyl diselenide

CAS: 1666-13-3; EC No: 216-780-2; MDL No: MFCD00003001; RTECS: JM9152500 UN No: UN3283; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: C12H10Se2; MW: 312.13 Melting Point: 59-61°


  • RTECS: JM9152500
  • Powder

Formula: C₁₂H₁₀Se₂
MW: 312.13 g/mol
Melting Pt: 59…61 °C
MDL Number: MFCD00003001
CAS Number: 1666-13-3
EINECS: 216-780-2
UN: 3283
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR