Potassium peroxodisulfate ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
di-Potassium peroxodisulphate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate
125925-1KG
125925-250G
BT125925-1KGEA
144.29
USD
BT125925-1KG
BT125925-250G
Potassium peroxodisulfate ≥99.0% ACS
Potassium peroxodisulphate
CAS: 7727-21-1; EC No: 231-781-8; MDL No: MFCD00011386; RTECS: SE0400000
UN No: UN1492; Haz Class: 5.1; Packing Group: III
Crystalline/Powder; Linear Formula: K2S2O8; MW: 270.31
Melting Point: 100°
Density (g/mL): 2.477
Moisture Sensitive
- RTECS: SE0400000
- Moisture Sensitive
- Crystalline/Powder
Formula:
K₂S₂O₈ MW: 270.32 g/mol Melting Pt: 100 °C Density: 2.477 g/mL |
MDL Number:
MFCD00011386 CAS Number: 7727-21-1 EINECS: 231-781-8 UN: 1492 ADR: 5.1,III |
Specification Test Results
Assay | 99.0% min. |
Iron (Fe) | ≤0.0005% |
Chloride (Cl) | ≤0.001% |
Manganese (Mn) | ≤0.0001% |
Heavy metals (as Pb) | ≤0.001% |
Insoluble matter | ≤0.003% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...