Dicyclohexylamine ≥98%
Supplier: BeanTown Chemical
CAS: 101-83-7; EC No: 202-980-7; MDL No: MFCD00011658; RTECS: HY4025000
UN No: UN2565; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C12H23N; MW: 181.32
Melting Point: -2°; Boiling Point: 256°; Flash point: 96°C (205°F)
Density (g/mL): 0.912; Refractive Index: 1.484
Air Sensitive
- RTECS: HY4025000
- Refractive index: 1.484
- Air Sensitive
- Liquid
Formula:
C₁₂H₂₃N Boiling Pt: 256 °C Melting Pt: –2 °C Density: 0.912 g/mL Flash Pt: 96 °C (205 °F) |
MDL Number:
MFCD00011658 CAS Number: 101-83-7 EINECS: 202-980-7 UN: 2565 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...