Aluminum silicon ≥99%, Powder -325 mesh

Supplier: BeanTown Chemical

125870-100G 125870-500G
BT125870-100GEA 65 USD
BT125870-100G BT125870-500G
Aluminum silicon ≥99%, Powder -325 mesh
Aluminum silicon

CAS: 11145-27-0; EC No: 231-072-3; MDL No: MFCD00143595 UN No: UN1398; Haz Class: 4.3; Packing Group: III Powder; Molecular Formula: Al:Si; 88:12 wt% ; MW: 55.07


  • Powder

Formula: Al:Si (88:12%)
MDL Number: MFCD00143595
CAS Number: 11145-27-0
EINECS: 231-072-3
ADR: 4.3,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR