Methyl trifluoroacetate ≥98%

Supplier: BeanTown Chemical

Synonyms: Trifluoroacetic methyl ester

125750-1KG 125750-250G 125750-50G
BT125750-1KGEA 907 USD
BT125750-1KG BT125750-250G BT125750-50G
Methyl trifluoroacetate ≥98%
Methyl trifluoroacetate

CAS: 431-47-0; EC No: 207-074-5; MDL No: MFCD00000417 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Linear Formula: CF3COOCH3; Molecular Formula: C3H3F3O2; MW: 128.05 Melting Point: -78°; Boiling Point: 43-44°; Flash point: -20°C (-4°F) Density (g/mL): 1.273; Refractive Index: 1.291 Moisture Sensitive


  • Refractive index: 1.291
  • Moisture Sensitive
  • Liquid

Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 43…44 °C
Melting Pt: –78 °C
Density: 1.273 g/mL
Flash Pt: –20 °C (–4 °F)
MDL Number: MFCD00000417
CAS Number: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR