Cobalt(II,III) oxide ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Tricobalt tetraoxide, Cobaltic tetraoxide
125740-1KG
125740-250G
125740-50G
BT125740-1KGEA
750
USD
BT125740-1KG
BT125740-250G
BT125740-50G
Cobalt(II,III) oxide ≥99.5% (trace metals basis)
Cobalt(II,III) oxide
CAS: 1308-06-1; EC No: 215-157-2; MDL No: MFCD00010939; RTECS: GG2500000
UN No: UN3077; Haz Class: 9; Packing Group: III
2-6 Micron Powder; Molecular Formula: Co3O4; MW: 240.80
Melting Point: 895° (decomposes)
Density (g/mL): 6.11
- RTECS: GG2500000
- 2-6 Micron Powder
Formula:
Co₃O₄ MW: 240.8 g/mol Melting Pt: 895 °C (dec.) Density: 6.11 g/mL |
MDL Number:
MFCD00010939 CAS Number: 1308-06-1 EINECS: 215-157-2 ADR: 9,III |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...