Strontium nitrate, anhydrous ≥98%

Supplier: BeanTown Chemical

125720-1KG 125720-250G
BT125720-1KGEA 76.28 USD
BT125720-1KG BT125720-250G
Strontium nitrate, anhydrous ≥98%
Strontium nitrate

CAS: 10042-76-9; EC No: 233-131-9; MDL No: MFCD00011248; RTECS: WK9800000 UN No: UN1507; Haz Class: 5.1; Packing Group: III Granular; Linear Formula: Sr(NO3)2; MW: 211.63 Melting Point: 570° Density (g/mL): 2.99 Hygroscopic


  • RTECS: WK9800000
  • Hygroscopic
  • Granular

Formula: Sr(NO₃)₂
MW: 211.63 g/mol
Boiling Pt: 645
Melting Pt: 570
Density: 2.99 g/mL
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1 III
Merck Index: 13,08926

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR