Triethyl phosphite ≥98%
Supplier: BeanTown Chemical
CAS: 122-52-1; EC No: 204-552-5; MDL No: MFCD00009084; RTECS: TH1130000
UN No: UN2323; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: (CH3CH2O)3P; Molecular Formula: C6H15O3P; MW: 166.16
Melting Point: -112°; Boiling Point: 155-157°; Flash point: 43°C (109°F)
Density (g/mL): 0.969; Refractive Index: 1.413
Air Sensitive, Moisture Sensitive
- RTECS: TH1130000
- Refractive index: 1.413
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₆H₁₅O₃P Boiling Pt: 155…157 °C Melting Pt: –112 °C Density: 0.969 g/mL Flash Pt: 43 °C (109 °F) |
MDL Number:
MFCD00009084 CAS Number: 122-52-1 EINECS: 204-552-5 UN: 2323 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...