Ethyl isothiocyanatoformate ≥97%

Supplier: BeanTown Chemical

Synonyms: Ethoxycarbonyl isothiocyanate

125685-25G 125685-5G
BT125685-25GEA 223 USD
BT125685-25G BT125685-5G
Ethyl isothiocyanatoformate ≥97%
Ethyl isothiocyanatoformate

CAS: 16182-04-0; EC No: 240-318-9; MDL No: MFCD00004814 UN No: UN2924; Haz Class: 3(8); Packing Group: III Oil; Linear Formula: SCNCOOCH2CH3; Molecular Formula: C4H5NO2S; MW: 131.15 Boiling Point: 56°/18 mmHg; Flash point: 50°C (122°F) Density (g/mL): 1.112; Refractive Index: 1.500 Moisture Sensitive


  • Refractive index: 1.500
  • Moisture Sensitive
  • Oil

Formula: C₄H₅NO₂S
MW: 131.16 g/mol
Boiling Pt: 56 °C/18 mmHg
Density: 1.112 g/mL
Flash Pt: 50 °C (122 °F)
MDL Number: MFCD00004814
CAS Number: 16182-04-0
EINECS: 240-318-9
ADR: 3(8),III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR