Methanol-[D₄] ≥99.8% (Isotopic)

Supplier: BeanTown Chemical

125660-10EACH 125660-2EACH
BT125660-10EAEA 148.4 USD
BT125660-10EA BT125660-2EACH
Methanol-[D₄] ≥99.8% (Isotopic)
Methanol-[D₄]

CAS: 811-98-3; EC No: 212-378-6; MDL No: MFCD00044637 UN No: UN1230; Haz Class: 3; Packing Group: II Liquid, in prescored ampules, 0.75mL/ampule; Linear Formula: CD3OD; MW: 36.07 Melting Point: -98°; Boiling Point: 65.4°; Flash point: 11°C (54°F) Density (g/mL): 0.888; Refractive Index: 1.326 Hygroscopic


  • Refractive index: 1.326
  • Hygroscopic
  • Liquid, in prescored ampules, 0.75mL/ampule

Formula: D₃COD
Boiling Pt: 65.4 °C
Melting Pt: –98 °C
Density: 0.888 g/mL
Flash Pt: 11 °C (54 °F)
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Assay 99.8%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR