Boron triiodide ≥98%

Supplier: BeanTown Chemical

125620-1G 125620-5G
BT125620-1GEA 66.2 USD
BT125620-1G BT125620-5G
Boron triiodide ≥98%
Boron triiodide

CAS: 13517-10-7; EC No: 236-857-4; MDL No: MFCD00036286 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: BI3; MW: 391.52 Density (g/mL): 3.35 Light Sensitive, Moisture Sensitive


  • Light Sensitive, Moisture Sensitive
  • Crystalline

Formula: BI₃
MW: 391.52 g/mol
Boiling Pt: 210
Melting Pt: 43
Density: 3.35 g/mL
MDL Number: MFCD00036286
CAS Number: 13517-10-7
EINECS: 236-857-4
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR