Carbazole ≥95%
Supplier: BeanTown Chemical
125550-100G
125550-500G
BT125550-100GEA
53.15
USD
BT125550-100G
BT125550-500G
Carbazole ≥95%
Carbazole
CAS: 86-74-8; EC No: 201-696-0; MDL No: MFCD00004960; RTECS: FE3150000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: C12H9N; MW: 167.21
Melting Point: 243-246°; Boiling Point: 355°; Flash point: 220°C (428°F)
- RTECS: FE3150000
- Powder
Formula:
C₁₂H₉N MW: 167.21 g/mol Boiling Pt: 355 °C Melting Pt: 243…246 °C Flash Pt: 220 °C (428 °F) |
MDL Number:
MFCD00004960 CAS Number: 86-74-8 EINECS: 201-696-0 ADR: 9,III |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...