4-Nitrophenol ≥99%
Supplier: BeanTown Chemical
125510-1KG
125510-250G
125510-5KG
BT125510-1KGEA
56.06
USD
BT125510-1KG
BT125510-250G
BT125510-5KG
4-Nitrophenol ≥99%
4-Nitrophenol
CAS: 100-02-7; EC No: 202-811-7; MDL No: MFCD00007331; RTECS: SM2275000
UN No: UN1663; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C6H5NO3; MW: 139.11
Melting Point: 110-115°; Boiling Point: 279°; Flash point: 169°C (336°F)
Light Sensitive
- RTECS: SM2275000
- Light Sensitive
- Crystalline
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Boiling Pt: 279 °C Melting Pt: 110…115 °C Flash Pt: 169 °C (336 °F) |
MDL Number:
MFCD00007331 CAS Number: 100-02-7 EINECS: 202-811-7 UN: 1663 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...