Barium perchlorate, anhydrous
Supplier: BeanTown Chemical
125465-100G
125465-500G
BT125465-100GEA
39
USD
BT125465-100G
BT125465-500G
Barium perchlorate, anhydrous
Barium perchlorate
CAS: 13465-95-7; EC No: 236-710-4; MDL No: MFCD00003440
UN No: UN1447; Haz Class: 5.1 (6.1); Packing Group: II
Crystalline; Linear Formula: Ba(ClO4)2; MW: 336.23
Melting Point: 505°
Density (g/mL): 3.2
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Ba(ClO₄)₂ MW: 336.23 g/mol Melting Pt: 505 °C Density: 3.2 g/mL |
MDL Number:
MFCD00003440 CAS Number: 13465-95-7 EINECS: 236-710-4 UN: 1447 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...