tert-Butyl bromoacetate ≥98%

Supplier: BeanTown Chemical

Synonyms: Bromoacetic tert-butyl ester

125460-10G 125460-250G 125460-50G
BT125460-10GEA 30.68 USD
BT125460-10G BT125460-250G BT125460-50G
tert-Butyl bromoacetate ≥98%
tert-Butyl bromoacetate

CAS: 5292-43-3; EC No: 226-133-6; MDL No: MFCD00000188 UN No: UN2924; Haz Class: 3(8); Packing Group: III Liquid; Linear Formula: (CH3)3COCOCH2Br; Molecular Formula: C6H11BrO2; MW: 195.06 Boiling Point: 50°/10 mmHg; Flash point: 49°C (120°F) Density (g/mL): 1.321; Refractive Index: 1.445


  • Refractive index: 1.445
  • Liquid

Formula: BrCH₂COOC(CH₃)₂
Boiling Pt: 50 °C/10 mmHg
Density: 1.321 g/mL
Flash Pt: 49 °C (120 °F)
MDL Number: MFCD00000188
CAS Number: 5292-43-3
EINECS: 226-133-6
ADR: 3(8),III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR