2,4-Xylenol ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Hydroxy-3-xylene, 2,4-Dimethylphenol, 2,4-Xylenol

125440-100G 125440-500G
BT125440-100GEA 38.73 USD
BT125440-100G BT125440-500G
2,4-Xylenol ≥98%
2,4-Xylenol

CAS: 105-67-9; EC No: 203-321-6; MDL No: MFCD00002233; RTECS: ZE5600000 UN No: UN2261; Haz Class: 6.1; Packing Group: II Liquid/Low Melting Solid; Molecular Formula: C8H10O; MW: 122.16 Melting Point: 22-23°; Boiling Point: 211-212°; Flash point: 94°C (201°F) Density (g/mL): 1.011; Refractive Index: 1.538


  • RTECS: ZE5600000
  • Refractive index: 1.538
  • Liquid/Low Melting Solid

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 211…212 °C
Melting Pt: 22…23 °C
Density: 1.011 g/mL
Flash Pt: 94 °C (201 °F)
MDL Number: MFCD00002233
CAS Number: 105-67-9
EINECS: 203-321-6
UN: 2261
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR