Trimethyl borate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

125425-100G 125425-25G 125425-500G
BT125425-100GEA 102 USD
BT125425-100G BT125425-25G BT125425-500G
Trimethyl borate ≥99.99% (trace metals basis)
Trimethyl borate

CAS: 121-43-7; EC No: 204-468-9; MDL No: MFCD00008346; RTECS: ED5600000 UN No: UN2416; Haz Class: 3; Packing Group: II Liquid; Linear Formula: B(OCH3)3; Molecular Formula: C3H9BO3; MW: 103.92 Melting Point: -34°; Boiling Point: 68-69°; Flash point: -8°C (18°F) Density (g/mL): 0.932; Refractive Index: 1.346 Moisture Sensitive


  • RTECS: ED5600000
  • Refractive index: 1.346
  • Moisture Sensitive
  • Liquid

Formula: C₃H₉BO₃
MW: 103.91 g/mol
Boiling Pt: 68…69 °C
Melting Pt: –34 °C
Density: 0.932 g/mL
Flash Pt: –8 °C (18 °F)
MDL Number: MFCD00008346
CAS Number: 121-43-7
EINECS: 204-468-9
UN: 2416
ADR: 3,II

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR