Lead(II) fluoride ≥99%

Supplier: BeanTown Chemical

Synonyms: Lead difluoride

125415-1KG 125415-250G 125415-50G
BT125415-1KGEA 282 USD
BT125415-1KG BT125415-250G BT125415-50G
Lead(II) fluoride ≥99%
Lead(II) fluoride

CAS: 7783-46-2; EC No: 231-998-8; MDL No: MFCD00011162 UN No: UN2291; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: PbF2; MW: 245.20 Melting Point: 824° Density (g/mL): 8.445


  • Powder

Formula: F₂Pb
MW: 245.2 g/mol
Melting Pt: 824 °C
Density: 8.445 g/mL
MDL Number: MFCD00011162
CAS Number: 7783-46-2
EINECS: 231-998-8
UN: 2291
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR