Potassium iodate 99.4-100.4% ACS

Supplier: BeanTown Chemical

125400-100G 125400-500G
BT125400-100GEA 55.62 USD
BT125400-100G BT125400-500G
Potassium iodate 99.4-100.4% ACS
Potassium iodate

CAS: 7758-05-6; EC No: 231-831-9; MDL No: MFCD00011406; RTECS: NN1350000 UN No: UN1479; Haz Class: 5.1; Packing Group: II Crystalline/Powder; Linear Formula: KIO3; MW: 214.00 Melting Point: 560° Density (g/mL): 3.93


  • RTECS: NN1350000
  • Crystalline/Powder

Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560 °C
Density: 3.93 g/mL
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.4-100.4%
pH of 5% solution at 25°C 5.0-8.0
Insoluble matter ≤0.005%
Chloride & bromide ≤0.01%
Iodine (I) ≤0.001%
Sulfate (SO4) ≤0.005%
Nitrogen compounds (as N) ≤0.005%
Sodium (Na) ≤0.005%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤0.001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR