Aluminium trifluoromethanesulphonate
Supplier: BeanTown Chemical
Synonyms:
Aluminum trifluoromethanesulphonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)
125390-10G
125390-50G
BT125390-10GEA
88
USD
BT125390-10G
BT125390-50G
Aluminium trifluoromethanesulphonate
Aluminium trifluoromethanesulphonate
CAS: 74974-61-1; MDL No: MFCD00143596
UN No: UN3261; Haz Class: 8; Packing Group: III
Powder; Linear Formula: Al(SO3CF3)3; Molecular Formula: C3AlF9O9S3; MW: 474.19
Melting Point: 300°
Hygroscopic
- Hygroscopic
- Powder
Formula:
C₃AlF₉O₉S₃ MW: 474.19 g/mol Melting Pt: 300 °C |
MDL Number:
MFCD00143596 CAS Number: 74974-61-1 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...