Aluminium chloride hexahydrate

Supplier: BeanTown Chemical

Synonyms: Aluminum chloride hexahydrate

125380-2KG 125380-500G
BT125380-2KGEA 179.19 USD
BT125380-2KG BT125380-500G
Aluminium chloride hexahydrate
Aluminium chloride hexahydrate

CAS: 7784-13-6; EC No: 231-208-1; MDL No: MFCD00149134; RTECS: BD0530000 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Molecular Formula: AlCl3·6H2O ; MW: 241.43


  • RTECS: BD0530000
  • Crystalline

Formula: AlCl₃·6H₂O
MW: 241.43 g/mol
Melting Pt: 100
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR