3,5-Dichlorophenol ≥98%

Supplier: BeanTown Chemical

125370-25G 125370-50G 125370-5G
BT125370-25GEA 204 USD
BT125370-25G BT125370-50G BT125370-5G
3,5-Dichlorophenol ≥98%
3,5-Dichlorophenol

CAS: 591-35-5; EC No: 209-714-9; MDL No: MFCD00002259; RTECS: SK8820000 UN No: UN2020; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Molecular Formula: C6H4Cl2O; MW: 163.00 Melting Point: 65-68°; Boiling Point: 233°


  • RTECS: SK8820000
  • Crystalline/Powder

Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 233 °C
Melting Pt: 65…68 °C
MDL Number: MFCD00002259
CAS Number: 591-35-5
EINECS: 209-714-9
UN: 2020
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR