(R)-(+)-3-Butyne-2-ol ≥99%

Supplier: BeanTown Chemical

Synonyms: (R)-(+)-But-3-yn-2-ol, (R)-But-3-yn-2-ol, (R)-(+)-3-Hydroxy-1-butyne

125355-1G 125355-250MG 125355-5G
BT125355-1GEA 170 USD
BT125355-1G BT125355-250MG BT125355-5G
(R)-(+)-3-Butyne-2-ol ≥99%
(R)-(+)-3-Butyne-2-ol

CAS: 42969-65-3; MDL No: MFCD00211237 UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II Liquid; Molecular Formula: C4H6O; MW: 70.09 Boiling Point: 108-111°; Flash point: 26°C (79°F) Density (g/mL): 0.89; Refractive Index: 1.426; Optical Rotation: [α]22/D +45°, neat


  • Optical rotation: [α]22/D +45°, neat
  • Refractive index: 1.426
  • Liquid

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 108…111 °C
Density: 0.89 g/mL
Flash Pt: 26 °C (79 °F)
MDL Number: MFCD00211237
CAS Number: 42969-65-3
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR