Silver nitrate 0.1 N in water volumetric solution

Supplier: BeanTown Chemical

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

125350-1L 125350-4L 125350-500ML
BT125350-1LEA 152.67 USD
BT125350-1L BT125350-4L BT125350-500ML
Silver nitrate 0.1 N in water volumetric solution
Silver nitrate

CAS: 7761-88-8; EC No: 231-853-9; MDL No: MFCD00003414 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Molecular Formula: AgNO3; MW: 169.87 Density (g/mL): 1.015 Light Sensitive


  • Liquid
  • Light Sensitive

Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 1.015 g/mL
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9 III
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR